5-Iodoindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20638-06
B20638-14
CAAAB20638-06EA
154.68
CAD
CAAAB20638-06
CAAAB20638-14
CAAAB20638-22
5-Iodoindole 98%
5-Iodoindole
Purity: 98%
CAS No.: 16066-91-4
Molecular Formula: C8H6IN
Formula Weight: 243.05
Melting Point: 97-100°
MDL No.: MFCD00220065
Formula:
C₈H₆IN Melting Pt: 97…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00220065 CAS Number: 16066-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...