(3,4-Dimethoxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20625-14 B20625-22 B20625-36
CAAAB20625-14EA 55.93 CAD
CAAAB20625-14 CAAAB20625-22 CAAAB20625-36
(3,4-Dimethoxyphenyl)acetic acid 98%
(3,4-Dimethoxyphenyl)acetic acid

Purity: 98% CAS No.: 93-40-3 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 95-98° Beilstein Reference: 1110282

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00004335
CAS Number: 93-40-3
EINECS: 202-244-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR