3,4,5-Trihydroxybenzamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20620-06
B20620-14
CAAAB20620-06EA
67.31
CAD
CAAAB20620-06
CAAAB20620-14
3,4,5-Trihydroxybenzamide 98%
3,4,5-Trihydroxybenzamide
Purity: 98%
CAS No.: 618-73-5
Molecular Formula: C7H7NO4
Formula Weight: 169.14
Melting Point: ca 240° dec.
Beilstein Reference: 2644549
Formula:
C₇H₇NO₄ Boiling Pt: 403 °C (1013 hPa) Melting Pt: ca 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00014800 CAS Number: 618-73-5 EINECS: 210-561-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...