2-Iodobutane 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
B20619-36
B20619-18
B20619-30
CAAAAB20619-36EA
461.36
CAD
CAAAAB20619-36
CAAAB20619-18
CAAAB20619-30
2-Iodobutane 99% stabilized
2-Iodobutane
Purity: 99%
CAS No.: 513-48-4
Molecular Formula: C4H9I
Formula Weight: 184.02
Flash Point: 21°(70°F)
Refractive Index: 1.4991
Odor: Ether-like
Melting Point: -104°
Boiling Point: 119-120°
Beilstein Reference: 1718777
Merck Reference: 14,1574
UN No.: 2390
MDL No.: MFCD00001072
Formula:
C₄H₉I Boiling Pt: 119…120 °C Melting Pt: –104 °C Density: 1.597 Flash Pt: 21 °C (70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001072 CAS Number: 513-48-4 EINECS: 208-163-1 UN: 2390 ADR: 3,II Merck Index: 14,1574 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...