4-Fluoro-2-(trifluoromethyl)benzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20617-14
B20617-06
CAAAAB20617-14EA
431.34
CAD
CAAAAB20617-14
CAAAB20617-03
CAAAB20617-06
4-Fluoro-2-(trifluoromethyl)benzonitrile 98+%
4-Fluoro-2-(trifluoromethyl)benzonitrile
Purity: 98+%
CAS No.: 194853-86-6
Molecular Formula: C8H3F4N
Formula Weight: 189.11
Melting Point: 43-45°
Boiling Point: 194-196°
UN No.: 3439
MDL No.: MFCD00061283
Formula:
C₈H₃F₄N Boiling Pt: 194…196 °C Melting Pt: 43…45 °C Density: 1.350 Storage Temperature: Ambient |
MDL Number:
MFCD00061283 CAS Number: 194853-86-6 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...