(±)-1-Hepten-4-ol ≥97%

Supplier: Thermo Scientific Chemicals
Warning

B20591-03 B20591-06 B20591-14
CAAAB20591-03EA 48.03 CAD
CAAAB20591-03 CAAAB20591-06 CAAAB20591-14
(±)-1-Hepten-4-ol ≥97%
(±)-1-Hepten-4-ol

Purity: 97% CAS No.: 3521-91-3 Molecular Formula: C7H14O Formula Weight: 114.18 UN No.: 1987 MDL No.: MFCD00021933

Formula: C₇H₁₄O
Boiling Pt: 152.1 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.8412 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00021933
CAS Number: 3521-91-3
EINECS: 222-535-0
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR