2,2,3,4,4,4-Hexafluoro-1-butanol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H,1H,3H-Perfluoro-1-butanol, 2,2,3,4,4,4-Hexafluorobutan-1-ol
B20587-22
B20587-06
B20587-14
CAAAAB20587-22EA
567.22
CAD
CAAAAB20587-22
CAAAB20587-06
CAAAB20587-14
2,2,3,4,4,4-Hexafluoro-1-butanol 95%
2,2,3,4,4,4-Hexafluoro-1-butanol
Purity: 95%
CAS No.: 382-31-0
Molecular Formula: CF3CHFCF2CH2OH
Formula Weight: 182.07
Flash Point: 51°(123°F)
Refractive Index: 1.3120
Boiling Point: 114-116°
Beilstein Reference: 1753872
UN No.: 1993
MDL No.: MFCD00042309
Formula:
C₄H₄F₆O MW: 182.07 g/mol Boiling Pt: 114…116 °C Density: 1.64 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042309 CAS Number: 382-31-0 EINECS: 206-842-7 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...