4-Ethoxyphenol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20584-09
B20584-18
CAAAB20584-09EA
47.56
CAD
CAAAB20584-09
CAAAB20584-18
4-Ethoxyphenol 99%
4-Ethoxyphenol
Purity: 99%
CAS No.: 622-62-8
Molecular Formula: C8H10O2
Formula Weight: 138.17
Odor: Phenol-like
Melting Point: 64-67°
Boiling Point: 121°/9mm
Beilstein Reference: 1907114
MDL No.: MFCD00002334
Formula:
C₈H₁₀O₂ Boiling Pt: 121 °C/9 mm Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002334 CAS Number: 622-62-8 EINECS: 210-748-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...