4'-Isopropylacetophenone 97%
Supplier: Thermo Scientific Chemicals
B20581-06
B20581-14
B20581-22
CAAAB20581-06EA
76.16
CAD
CAAAB20581-06
CAAAB20581-14
CAAAB20581-22
4'-Isopropylacetophenone 97%
4'-Isopropylacetophenone
Purity: 97%
CAS No.: 645-13-6
Molecular Formula: C11H14O
Formula Weight: 162.23
Refractive Index: 1.5230
Boiling Point: 238°
MDL No.: MFCD00048297
Formula:
C₁₁H₁₄O Boiling Pt: 238 °C Density: 0.970 Storage Temperature: Ambient |
MDL Number:
MFCD00048297 CAS Number: 645-13-6 EINECS: 211-433-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...