1-Butylcyclohexanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-n-Butylcyclohexanol

B20579-06 B20579-14
CAAAB20579-06EA 107.76 CAD
CAAAB20579-06 CAAAB20579-14
1-Butylcyclohexanol 98%
1-Butylcyclohexanol

Purity: 98% CAS No.: 5445-30-7 Molecular Formula: C10H20O Formula Weight: 156.27 MDL No.: MFCD00021404

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021404
CAS Number: 5445-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR