3-Amino-5-tert-butylisoxazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20567-06
B20567-14
B20567-22
CAAAB20567-06EA
98.75
CAD
CAAAB20567-06
CAAAB20567-14
CAAAB20567-22
3-Amino-5-tert-butylisoxazole 97%
3-Amino-5-tert-butylisoxazole
Purity: 97%
CAS No.: 55809-36-4
Molecular Formula: C7H12N2O
Formula Weight: 140.19
Melting Point: 110-113°
Beilstein Reference: 1100990
Formula:
C₇H₁₂N₂O Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055620 CAS Number: 55809-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...