Toluene-2-sulfonamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20544-09
B20544-18
CAAAB20544-09EA
64.78
CAD
CAAAB20544-09
CAAAB20544-18
Toluene-2-sulfonamide 99%
Toluene-2-sulphonamide
Purity: 99%
CAS No.: 88-19-7
Molecular Formula: C7H9NO2S
Formula Weight: 171.22
Flash Point: 178°(352°F)
Melting Point: 156°
Boiling Point: 210°/1mm
Beilstein Reference: 1102362
MDL No.: MFCD00007934
Formula:
C₇H₉NO₂S Boiling Pt: 210 °C/1 mm Melting Pt: 156 °C Flash Pt: 178 °C(352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007934 CAS Number: 88-19-7 EINECS: 201-808-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...