2,2-Difluoro-1,3-benzodioxole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20540-06
B20540-14
CAAAB20540-06EA
114.23
CAD
CAAAB20540-06
CAAAB20540-14
2,2-Difluoro-1,3-benzodioxole 97%
2,2-Difluoro-1,3-benzodioxole
Purity: 97%
CAS No.: 1583-59-1
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Flash Point: 32°(90°F)
Refractive Index: 1.4430
Boiling Point: 129-130°
Beilstein Reference: 1307648
UN No.: 1993
MDL No.: MFCD00236217
Formula:
C₇H₄F₂O₂ Boiling Pt: 129…130 °C Density: 1.308 Flash Pt: 32 °C (90 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00236217 CAS Number: 1583-59-1 EINECS: 216-431-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...