2-Fluoro-3-methylbenzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Bromomethyl)-2-fluoro-3-methylbenzene
B20538-03
B20538-06
CAAAAB20538-14EA
3649.75
CAD
CAAAAB20538-14
CAAAB20538-03
CAAAB20538-06
2-Fluoro-3-methylbenzyl bromide 97%
2-Fluoro-3-methylbenzyl bromide
Purity: 97%
CAS No.: 151412-12-3
Molecular Formula: C8H8BrF
Formula Weight: 203.06
Refractive Index: 1.5480
Boiling Point: 105-106°/10mm
UN No.: 3265
MDL No.: MFCD00042470
Formula:
C₈H₈BrF Boiling Pt: 105…106 °C/10 mm Storage Temperature: Ambient |
MDL Number:
MFCD00042470 CAS Number: 151412-12-3 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...