N-Methylanthranilic acid 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20531-14
B20531-22
B20531-36
CAAAAB20531-14EA
62.09
CAD
CAAAAB20531-14
CAAAB20531-22
CAAAB20531-36
N-Methylanthranilic acid 90+%
N-Methylanthranilic acid
Purity: 90+%
CAS No.: 119-68-6
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: ca 171° dec.
Beilstein Reference: 2208135
Formula:
C₈H₉NO₂ Boiling Pt: 80 °C (0.01 -rr) Melting Pt: ca 171 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002424 CAS Number: 119-68-6 EINECS: 204-343-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...