2,6-Dichloro-3-nitrotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20520-09
B20520-18
CAAAAB20520-06EA
58.49
CAD
CAAAAB20520-06
CAAAB20520-09
CAAAB20520-18
2,6-Dichloro-3-nitrotoluene 99%
2,6-Dichloro-3-nitrotoluene
Purity: 99%
CAS No.: 29682-46-0
Molecular Formula: C7H5Cl2NO2
Formula Weight: 206.03
Melting Point: 53-55°
Beilstein Reference: 2259855
UN No.: 2811
MDL No.: MFCD00024185
Formula:
C₇H₅Cl₂NO₂ Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024185 CAS Number: 29682-46-0 EINECS: 249-776-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...