3-Ethoxy-4-methoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20489-03 B20489-06
CAAAB20489-03EA 57.99 CAD
CAAAB20489-03 CAAAB20489-06
3-Ethoxy-4-methoxybenzoic acid 98%
3-Ethoxy-4-methoxybenzoic acid

Purity: 98% CAS No.: 2651-55-0 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 156-160° MDL No.: MFCD00276591

Formula: C₁₀H₁₂O₃
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00276591
CAS Number: 2651-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR