2-Fluoro-4-(trifluoromethyl)benzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Cyano-3-fluorobenzotrifluoride
B20485-14
B20485-03
B20485-06
CAAAAB20485-14EA
1252.94
CAD
CAAAAB20485-14
CAAAB20485-03
CAAAB20485-06
2-Fluoro-4-(trifluoromethyl)benzonitrile 98%
2-Fluoro-4-(trifluoromethyl)benzonitrile
Purity: 98%
CAS No.: 146070-34-0
Molecular Formula: C8H3F4N
Formula Weight: 189.11
Flash Point: 75°(167°F)
Refractive Index: 1.4460
Boiling Point: 178-180°
Beilstein Reference: 8313354
UN No.: 3276
MDL No.: MFCD00061279
Formula:
C₈H₃F₄N MW: 189.11 Boiling Pt: 178…180 °C Density: 1.35 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061279 CAS Number: 146070-34-0 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...