2-Methoxy-1-naphthaldehyde 99%
Supplier: Thermo Scientific Chemicals
B20483-06
B20483-14
CAAAB20483-06EA
39.5
CAD
CAAAB20483-06
CAAAB20483-14
CAAAB20483-22
2-Methoxy-1-naphthaldehyde 99%
2-Methoxy-1-naphthaldehyde
Purity: 99%
CAS No.: 5392-12-1
Molecular Formula: C12H10O2
Formula Weight: 186.21
Melting Point: 81-85°
Boiling Point: 204-206°/18mm
MDL No.: MFCD00004004
Formula:
C₁₂H₁₀O₂ Boiling Pt: 204…206 °C/18 mm Melting Pt: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004004 CAS Number: 5392-12-1 EINECS: 226-392-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...