1-Bromo-5-chloropentane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20479-09
B20479-18
CAAAB20479-09EA
72.21
CAD
CAAAB20479-09
CAAAB20479-18
CAAAB20479-30
1-Bromo-5-chloropentane 98%
1-Bromo-5-chloropentane
Purity: 98%
CAS No.: 54512-75-3
Molecular Formula: Cl(CH2)5Br
Formula Weight: 185.50
Flash Point: 95°(203°F)
Refractive Index: 1.4836
Boiling Point: 210-212°
Beilstein Reference: 1732341
MDL No.: MFCD00001016
Formula:
BrH₂C(CH₂)₃CH₂Cl Boiling Pt: 210…212 °C Density: 1.408 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001016 CAS Number: 54512-75-3 EINECS: 259-193-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...