Tetrafluorosuccinic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

B20462-03 B20462-06
CAAAB20462-03EA 35.87 CAD
CAAAB20462-03 CAAAB20462-06
Tetrafluorosuccinic acid 97%
Tetrafluorosuccinic acid

Purity: 97% CAS No.: 377-38-8 Molecular Formula: C4H2F4O4 Formula Weight: 190.05 Melting Point: ca 115° Boiling Point: 150°/15mm UN No.: 3261 MDL No.: MFCD00042430

Formula: C₄H₂F₄O₄
Boiling Pt: 150 °C/15 mm
Melting Pt: ∼ 115 °C
Density: 1.400
Storage Temperature: Ambient
MDL Number: MFCD00042430
CAS Number: 377-38-8
EINECS: 206-820-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR