2-Chlorobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20451-0B
B20451-30
CAAAB20451-0BEA
297.04
CAD
CAAAB20451-0B
CAAAB20451-30
2-Chlorobenzonitrile 98%
2-Chlorobenzonitrile
Purity: 98%
CAS No.: 873-32-5
Molecular Formula: C7H4ClN
Formula Weight: 137.57
Flash Point: 122°(252°F)
Odor: Almond-like
Boiling Point: 232°
Beilstein Reference: 1363916
UN No.: 3439
MDL No.: MFCD00001779
Formula:
C₇H₄ClN Boiling Pt: 232 °C Melting Pt: 43…46 °C Density: 1.19 g/cm³ (25 °C) Flash Pt: 122 °C(252 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001779 CAS Number: 873-32-5 EINECS: 212-836-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...