1,4-Cyclohexanediol cis- and trans-mixture 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cyclohexane-1,4-diol
B20446-14
B20446-22
CAAAB20446-14EA
39.82
CAD
CAAAB20446-14
CAAAB20446-22
1,4-Cyclohexanediol cis- and trans-mixture 98%
1,4-Cyclohexanediol
Purity: 98%
CAS No.: 556-48-9
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 65°(149°F)
Melting Point: 98-103°
Boiling Point: 149-151°/20mm
Beilstein Reference: 2036658
MDL No.: MFCD00001448
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 149…151 °C/20 mm Melting Pt: 98…103 °C Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001448 CAS Number: 556-48-9 EINECS: 209-126-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...