(3-Chloropropyl)benzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B20445-06 B20445-14 B20445-22
CAAAB20445-06EA 41.4 CAD
CAAAB20445-06 CAAAB20445-14 CAAAB20445-22
(3-Chloropropyl)benzene 98%
(3-Chloropropyl)benzene

Purity: 98% CAS No.: 104-52-9 Molecular Formula: C9H11Cl Formula Weight: 154.64 Flash Point: 87°(189°F) Refractive Index: 1.5220 Boiling Point: 218-220° MDL No.: MFCD00001001

Formula: ClH₂CCH₂CH₂(C₆H₅)
Boiling Pt: 218…220 °C
Melting Pt: –27 °C
Density: 1.05
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001001
CAS Number: 104-52-9
EINECS: 203-210-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR