(2,4-Difluorophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Difluorophenylacetic acid

B20431-14 B20431-06
CAAAAB20431-14EA 736.28 CAD
CAAAAB20431-14 CAAAB20431-06
(2,4-Difluorophenyl)acetic acid 99%
(2,4-Difluorophenyl)acetic acid

Purity: 99% CAS No.: 81228-09-3 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 117-119° Beilstein Reference: 3649727

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00009999
CAS Number: 81228-09-3
EINECS: 279-709-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR