3,3,3-Trifluoropropionic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

B20428-14 B20428-06
CAAAAB20428-14EA 657.28 CAD
CAAAAB20428-14 CAAAB20428-06
3,3,3-Trifluoropropionic acid 98%
3,3,3-Trifluoropropionic acid

Purity: 98% CAS No.: 2516-99-6 Molecular Formula: CF3CH2CO2H Formula Weight: 128.05 Flash Point: 100°(212°F) Refractive Index: 1.3340 Melting Point: 9° Boiling Point: 146° Beilstein Reference: 1751796 UN No.: 3265 MDL No.: MFCD00153292

Formula: C₃H₃F₃O₂
Boiling Pt: 146 °C
Melting Pt: 9 °C
Density: 1.441
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR