3,5-Dinitrosalicylaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20424-03
B20424-06
CAAAB20424-03EA
83.74
CAD
CAAAB20424-03
CAAAB20424-06
3,5-Dinitrosalicylaldehyde 98%
3,5-Dinitrosalicylaldehyde
Purity: 98%
CAS No.: 2460-59-5
Molecular Formula: C7H4N2O6
Formula Weight: 212.12
Melting Point: 71-74°
Beilstein Reference: 1121900
Formula:
C₇H₄N₂O₆ MW: 212.12 g/mol Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007103 CAS Number: 2460-59-5 EINECS: 219-551-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...