4-Nitrobenzophenone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Nitrobenzophenone

B20405-06 B20405-14
CAAAB20405-06EA 49.77 CAD
CAAAB20405-06 CAAAB20405-14 CAAAB20405-22
4-Nitrobenzophenone 99%
4-Nitrobenzophenone

Purity: 99% CAS No.: 1144-74-7 Molecular Formula: C13H9NO3 Formula Weight: 227.22 Melting Point: 136-138° Beilstein Reference: 1912753

Formula: C₁₃H₉NO₃
Melting Pt: 136…138 °C
Density: 1.406 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007354
CAS Number: 1144-74-7
EINECS: 214-542-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR