Mercaptoacetic acid (thioglycolic acid) 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Reagent that protects tryptophan in amino acid analysis
Purity: 97+%
CAS No.: 68-11-1
Molecular Formula: C2H4O2S
Formula Weight: 92.11
Flash Point: 119°(246°F)
Physical Form: Clear liquid
Density: 1.295
Refractive Index: 1.5045
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Stench
Melting Point: -12°
Boiling Point: 220°
Beilstein Reference: 506166
Merck Reference: 14,9336
UN No.: 1940
MDL No.: MFCD00004876
Reagent that protects tryptophan in amino acid analysis
Formula:
HSCH₂COOH Boiling Pt: 220 °C Melting Pt: –12 °C Density: 1.295 Flash Pt: 119 °C (246 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004876 CAS Number: 68-11-1 EINECS: 200-677-4 UN: 1940 ADR: 8,II Merck Index: 14,9336 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...