4-(Dimethylamino)benzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

B20389-06 B20389-14 B20389-22
CAAAB20389-06EA 77.26 CAD
CAAAB20389-06 CAAAB20389-14 CAAAB20389-22
4-(Dimethylamino)benzophenone 98%
4-(Dimethylamino)benzophenone

Purity: 98% CAS No.: 530-44-9 Molecular Formula: C15H15NO Formula Weight: 225.29 Melting Point: 88-90° Merck Reference: 14,3233 MDL No.: MFCD00008311

Formula: C₁₅H₁₅NO
MW: 225.29 g/mol
Boiling Pt: 245 - 250 °C (15 -rr)
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008311
CAS Number: 530-44-9
EINECS: 208-478-4
Merck Index: 14,3233

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR