3,3-Dimethylglutaric acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

B20388-30 B20388-09 B20388-18
CAAAAB20388-30EA 627.26 CAD
CAAAAB20388-30 CAAAB20388-09 CAAAB20388-18
3,3-Dimethylglutaric acid 98+%
3,3-Dimethylglutaric acid

Purity: 98+% CAS No.: 4839-46-7 Molecular Formula: C7H12O4 Formula Weight: 160.17 Melting Point: 98-103° Boiling Point: 89-90°/2mm MDL No.: MFCD00002716

Formula: C₇H₁₂O₄
Boiling Pt: 89…90 °C/2 mm
Melting Pt: 98…103 °C
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00002716
CAS Number: 4839-46-7
EINECS: 225-425-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR