2,3,4-Trihydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B20371-14 B20371-03 B20371-06
CAAAAB20371-14EA 489.8 CAD
CAAAAB20371-14 CAAAB20371-03 CAAAB20371-06
2,3,4-Trihydroxybenzoic acid 97%
2,3,4-Trihydroxybenzoic acid

Purity: 97% CAS No.: 610-02-6 Molecular Formula: C7H6O5 Formula Weight: 170.12 Melting Point: ca 210° dec. Beilstein Reference: 2210807

Formula: C₇H₆O₅
MW: 170.12 g/mol
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002447
CAS Number: 610-02-6
EINECS: 210-205-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR