2-Amino-2',5-dichlorobenzophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20360-06
B20360-09
B20360-18
CAAAAB20360-06EA
48.19
CAD
CAAAAB20360-06
CAAAB20360-09
CAAAB20360-18
2-Amino-2',5-dichlorobenzophenone 99%
2-Amino-2',5-dichlorobenzophenone
Purity: 99%
CAS No.: 2958-36-3
Molecular Formula: C13H9Cl2NO
Formula Weight: 266.13
Odor: Odorless
Melting Point: 87-89°
Beilstein Reference: 652774
MDL No.: MFCD00007840
Formula:
C₁₃H₉Cl₂NO Boiling Pt: 385 °C (1013 hPa) Melting Pt: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007840 CAS Number: 2958-36-3 EINECS: 220-985-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...