1,2-Dibromobutane 98%

Supplier: Thermo Scientific Chemicals
Warning

B20356-18 B20356-30
CAAAAB20356-0BEA 895.51 CAD
CAAAAB20356-0B CAAAB20356-18 CAAAB20356-30
1,2-Dibromobutane 98%
1,2-Dibromobutane

Purity: 98% CAS No.: 533-98-2 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: 110°(230°F) Refractive Index: 1.5141 Melting Point: -65° Boiling Point: 166-167° Beilstein Reference: 1731380 Merck Reference: 14,1566 MDL No.: MFCD00000157

Formula: BrH₂CBrCHCH₂CH₃
Boiling Pt: 166…167 °C
Melting Pt: –65 °C
Density: 1.789
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000157
CAS Number: 533-98-2
EINECS: 208-581-4
UN: 2810
ADR: 6.1,III
Merck Index: 14,1566

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR