2,5-Dimethyl-2-hexanol 98%

Supplier: Thermo Scientific Chemicals
Warning

B20352-06 B20352-14
CAAAB20352-06EA 87.37 CAD
CAAAB20352-06 CAAAB20352-14 CAAAB20352-22
2,5-Dimethyl-2-hexanol 98%
2,5-Dimethyl-2-hexanol

Purity: 98% CAS No.: 3730-60-7 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 52°(125°F) Refractive Index: 1.4220 Boiling Point: 155-156° Beilstein Reference: 1732761 UN No.: 1987 MDL No.: MFCD00021814

Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 152
Flash Pt: 52 °C (125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021814
CAS Number: 3730-60-7
EINECS: 223-085-8
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR