(Trifluoromethyl)trimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trimethyl(trifluoromethyl)silane

B20347-14 B20347-03 B20347-06
CAAAAB20347-14EA 848.46 CAD
CAAAAB20347-14 CAAAB20347-03 CAAAB20347-06
(Trifluoromethyl)trimethylsilane 98%
(Trifluoromethyl)trimethylsilane

Purity: 98% CAS No.: 81290-20-2 Molecular Formula: C4H9F3Si Formula Weight: 142.20 Flash Point: -10°(14°F) Refractive Index: 1.3305 Boiling Point: 54-55° Beilstein Reference: 4241868 UN No.: 1993 MDL No.: MFCD00145454

Formula: C₄H₉F₃Si
MW: 142.2
Boiling Pt: 55.5
Density: 0.962
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR