4-Nitrothioanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

B20339-06 B20339-14
CAAAB20339-06EA 61.78 CAD
CAAAB20339-06 CAAAB20339-14 CAAAB20339-22
4-Nitrothioanisole 98%
4-Nitrothioanisole

Purity: 98% CAS No.: 701-57-5 Molecular Formula: C7H7NO2S Formula Weight: 169.20 Melting Point: 66-72° Beilstein Reference: 1817928

Formula: C₇H₇NO₂S
Boiling Pt: 135 - 140 °C (2 -rr)
Melting Pt: 66…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00010868
CAS Number: 701-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR