4-Methylthiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylbenzothioamide, Thio-p-toluamide

B20320-03 B20320-06
CAAAB20320-03EA 42.82 CAD
CAAAB20320-03 CAAAB20320-06
4-Methylthiobenzamide 97%
4-Methylthiobenzamide

Purity: 97% CAS No.: 2362-62-1 Molecular Formula: C8H9NS Formula Weight: 151.23 Melting Point: 166-169° MDL No.: MFCD00173750

Formula: C₈H₉NS
MW: 151.23 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00173750
CAS Number: 2362-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR