5-Bromoindole 99%
Supplier: Thermo Scientific Chemicals
B20307-06
B20307-14
B20307-22
CAAAB20307-06EA
53.25
CAD
CAAAB20307-06
CAAAB20307-14
CAAAB20307-22
5-Bromoindole 99%
5-Bromoindole
Purity: 99%
CAS No.: 10075-50-0
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Odor: Odorless
Melting Point: 89-92°
Beilstein Reference: 112877
MDL No.: MFCD00005670
Formula:
C₈H₆BrN Melting Pt: 89…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005670 CAS Number: 10075-50-0 EINECS: 233-208-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...