3',5'-Difluoroacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(3,5-Difluorophenyl)ethan-1-one
B20301-14
B20301-03
CAAAAB20301-14EA
1014.36
CAD
CAAAAB20301-14
CAAAB20301-03
CAAAB20301-06
3',5'-Difluoroacetophenone 97%
3',5'-Difluoroacetophenone
Purity: 97%
CAS No.: 123577-99-1
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Flash Point: 82°(179°F)
Refractive Index: 1.4876
Melting Point: 37-41°
Boiling Point: 94-95°/13mm
Beilstein Reference: 6798740
MDL No.: MFCD00042489
Formula:
C₈H₆F₂O MW: 156.13 g/mol Boiling Pt: 94…95 °C/13 mm Melting Pt: 37…41 °C Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042489 CAS Number: 123577-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...