Orcein for analysis

Supplier: Thermo Scientific Chemicals
Warning

B20294-03 B20294-06
CAAAB20294-03EA 60.36 CAD
CAAAB20294-03 CAAAB20294-06
Orcein for analysis
Orcein

Red in acidic pH and blue in alkaline pH


CAS No.: 1400-62-0 Merck Reference: 14,6863 MDL No.: MFCD00062310

Storage Temperature: Ambient
MDL Number: MFCD00062310
CAS Number: 1400-62-0
EINECS: 215-750-6
Merck Index: 14,6863

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR