1,3-Di-tert-butylbenzene 99%
Supplier: Thermo Scientific Chemicals
Warning
|
B20290-03
B20290-06
B20290-14
CAAAB20290-03EA
139.83
CAD
CAAAB20290-03
CAAAB20290-06
CAAAB20290-14
1,3-Di-tert-butylbenzene 99%
1,3-Di-tert-butylbenzene
Purity: 99%
CAS No.: 1014-60-4
Molecular Formula: C14H22
Formula Weight: 190.33
Flash Point: 83°(183°F)
Refractive Index: 1.4880
Melting Point: 10-11°
Boiling Point: 106-107°/18mm
MDL No.: MFCD00008830
Formula:
C₁₄H₂₂ Boiling Pt: 106…107 °C/18 mm Melting Pt: 10…11 °C Density: 0.859 Flash Pt: 83 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008830 CAS Number: 1014-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...