2',4'-Dihydroxy-3'-methylacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2,4-Dihydroxy-3-methylphenyl)ethanone, 1-(2,4-Dihydroxy-3-methylphenyl)ethan-1-one

B20272-06 B20272-14
CAAAB20272-06EA 102.38 CAD
CAAAB20272-06 CAAAB20272-14
2',4'-Dihydroxy-3'-methylacetophenone 98%
2',4'-Dihydroxy-3'-methylacetophenone

Purity: 98% CAS No.: 10139-84-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 152-156° MDL No.: MFCD00010817

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00010817
CAS Number: 10139-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR