2-Ethyl-1,3-cyclopentanedione 98+%
Supplier: Thermo Scientific Chemicals
B20254-06
B20254-14
CAAAB20254-06EA
63.83
CAD
CAAAB20254-06
CAAAB20254-14
CAAAB20254-22
2-Ethyl-1,3-cyclopentanedione 98+%
2-Ethyl-1,3-cyclopentanedione
Purity: 98+%
CAS No.: 823-36-9
Molecular Formula: C7H10O2
Formula Weight: 126.16
Storage Temperature: -30°C to -10°C
Melting Point: 168-170°
Beilstein Reference: 1860068
MDL No.: MFCD00044191
Formula:
C₇H₁₀O₂ Melting Pt: 168…170 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00044191 CAS Number: 823-36-9 EINECS: 212-512-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...