1-(2-Hydroxyphenyl)piperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20252-09
B20252-18
CAAAB20252-09EA
97.8
CAD
CAAAB20252-09
CAAAB20252-18
1-(2-Hydroxyphenyl)piperazine 98%
1-(2-Hydroxyphenyl)piperazine
Purity: 98%
CAS No.: 1011-17-2
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 122-127°
Beilstein Reference: 10646
UN No.: 3263
MDL No.: MFCD00190246
Formula:
C₁₀H₁₄N₂O Boiling Pt: 318 °C (1013 hPa) Melting Pt: 122…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190246 CAS Number: 1011-17-2 EINECS: 213-782-5 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...