Methyl-N-methylanthranilate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20233-18
B20233-30
CAAAB20233-18EA
65.41
CAD
CAAAB20233-18
CAAAB20233-30
Methyl-N-methylanthranilate 98%
Methyl N-methylanthranilate
Purity: 98%
CAS No.: 85-91-6
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 133°(271°F)
Refractive Index: 1.5800
Melting Point: 17-19°
Boiling Point: 256-258°
Beilstein Reference: 607217
MDL No.: MFCD00017183
Formula:
C₉H₁₁NO₂ Boiling Pt: 256…258 °C Melting Pt: 17…19 °C Density: 1.130 Flash Pt: 133 °C(271 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017183 CAS Number: 85-91-6 EINECS: 201-642-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...