(±)-3-Hexanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-Propyl ethyl carbinol, (±)-Ethyl propyl carbinol, (±)-3-Hydroxy hexane
B20210-14
B20210-22
CAAAB20210-14EA
124.03
CAD
CAAAB20210-14
CAAAB20210-22
(±)-3-Hexanol 98%
(±)-3-Hexanol
Purity: 98%
CAS No.: 623-37-0
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 42°(108°F)
Refractive Index: 1.4150
Odor: Characteristic
Boiling Point: 134-136°
Beilstein Reference: 1718964
UN No.: 2282
MDL No.: MFCD00004582
Formula:
C₆H₁₄O Boiling Pt: 134…136 °C Melting Pt: –70 °C Density: 0.81 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004582 CAS Number: 623-37-0 EINECS: 210-790-0 UN: 2282 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...