2,4-Difluorophenyl isothiocyanate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20204-03
B20204-06
CAAAB20204-03EA
47.08
CAD
CAAAB20204-03
CAAAB20204-06
2,4-Difluorophenyl isothiocyanate 96%
2,4-Difluorophenyl isothiocyanate
Purity: 96%
CAS No.: 141106-52-7
Molecular Formula: C7H3F2NS
Formula Weight: 171.17
Flash Point: 92°(197°F)
Refractive Index: 1.5950
Boiling Point: 98-99°/30mm
Beilstein Reference: 6919744
UN No.: 2922
MDL No.: MFCD00041044
Formula:
C₇H₃F₂NS Boiling Pt: 98…99 °C/30 mm Density: 1.349 Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041044 CAS Number: 141106-52-7 EINECS: 624-035-1 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...