(3-Bromophenyl)acetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20189-22
B20189-06
B20189-14
CAAAAB20189-22EA
725.22
CAD
CAAAAB20189-22
CAAAB20189-06
CAAAB20189-14
(3-Bromophenyl)acetonitrile 97%
(3-Bromophenyl)acetonitrile
Purity: 97%
CAS No.: 31938-07-5
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Flash Point: 126°(259°F)
Refractive Index: 1.5700
Melting Point: 25-28°
Boiling Point: 275°
Beilstein Reference: 2355439
UN No.: 3449
MDL No.: MFCD00001906
Formula:
C₈H₆BrN Boiling Pt: 275 °C Melting Pt: 25…28 °C Flash Pt: 126 °C (259 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001906 CAS Number: 31938-07-5 EINECS: 250-867-6 UN: 3449 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...