N,N-Dimethylethylendiamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

B20182-22 B20182-18 B20182-30
CAAAAB20182-22EA 158 CAD
CAAAAB20182-22 CAAAB20182-18 CAAAB20182-30
N,N-Dimethylethylendiamine 97%
N,N-Dimethylethylendiamine

Purity: 97% CAS No.: 108-00-9 Molecular Formula: C4H12N2 Formula Weight: 88.15 Flash Point: 12°(53°F) Refractive Index: 1.4270 Melting Point: -70° Boiling Point: 107° Beilstein Reference: 605279 UN No.: 2733 MDL No.: MFCD00008175

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 107 °C
Melting Pt: < –70 °C
Density: 0.810
Flash Pt: 12 °C(53 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR